missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric acid concentrate, For 1L standard solution, 0.1 M HClO4 (0.1N), for titration in non-aqueous medium, Honeywell Fluka™
Click to view available options
Quantity:
1 L
6 x 1 Ea.
Packaging:
Ampoule
Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Packaging | Bottle |
Flash Point | 113°C (235.4°F) |
Quantity | 1 L |
Linear Formula | HClO4 |
UN Number | 1873 |
Formula Weight | 100.46g/mol |
Chemical Name or Material | Perchloric Acid Concentrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'