Learn More
Perchloric acid, ACS, 48-50%, Thermo Scientific Chemicals
Description
Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 112°C |
Density | 1.41 g/cm3 at 20°C |
Vapor Pressure | 29mm Hg at 20°C |
Assay Percent Range | 48-50% |
Quantity | 500 mL |
Physical Form | Liquid |
UN Number | UN1802 |
Solubility Information | Miscible with water. |
Formula Weight | 100.46 |
Grade | ACS |
Show More |
RUO – Research Use Only