missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 60%, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073371
Click to view available options
Quantity:
500 mL
2.5 L
7601-90-3
ClHO4
100.45
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
24247
CHEBI:29221
perchloric acid
O[Cl](=O)(=O)=O
7601-90-3
ClHO4
100.45
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
24247
CHEBI:29221
perchloric acid
O[Cl](=O)(=O)=O
pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Packaging Plastic Coated Glass bottle
Vapor Pressure 6.8mmHg 25
Quantity 500 mL
Viscosity 3.5 mPaS at 20°C
Physical Form Liquid
Formula Weight 100.46g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'