missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 60%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15653270 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 15653270

Brand: Honeywell-Fluka 3114131L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent, 60%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Melting Point -18°C
Boiling Point 203°C
Density 1.535g/mL at 25°C
Packaging Glass Bottle
Residue after Ignition ≤0.003%
Vapor Pressure 7mmHg (25°C)
Flash Point 113°C (235.4°F)
Quantity 1 L
Linear Formula HClO4
UN Number UN1873
Formula Weight 100.46g/mol
Percent Purity 60%
Grade ACS Reagent
Chemical Name or Material Perchloric Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.