missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peracetic Acid Solution, purum, Honeywell™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Specifications
Density | 1.15g/mL at 20°C |
Packaging | Glass bottle |
Linear Formula | CH3CO3H |
Quantity | 100 mL |
Beilstein | 1098464 |
Formula Weight | 76.05g/mol |
Grade | purum p.a. |
Concentration or Composition (by Analyte or Components) | 39% in acetic acid |
Chemical Name or Material | Peracetic acid solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'