missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PER1 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | PER1 |
---|---|
Concentration | LYOPH |
Dilution | Immunohistochemistry 5-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Description
PER1 Polyclonal specifically detects PER1 in Human samples. It is validated for Immunohistochemistry.Specifications
PER1 | |
Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Circadian clock protein PERIOD 1, Circadian pacemaker protein Rigui, hPER1, KIAA0482, PERhPER, period (Drosophila) homolog 1, period circadian protein homolog 1, period homolog 1 (Drosophila), Period, drosophila, homolog of, PERIOD1, RIGUI, RIGUIMGC88021 | |
PER1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
O15534 | |
5187 | |
E. coli-derived recombinant human PER1 Ser159-Thr365 Accession # O15534 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human PER1 in direct ELISAs. |
For Research Use Only