missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PepSwift™ Monolithic Capillary LC Columns

For fast, high-resolution LC/MS separations in protein identification, biomarker discovery, and systems biology applications use these monolithic columns.

Brand:  Thermo Scientific™ 164584

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11382263

  • £795.00 / Each


Estimated Shipment
12-02-2018
Add to basket

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Specifications

Analytical Column, NanoViper
0.1mm
50mm
4350psi
Monolith
80°C
Phenyl
Reversed Phase
700 to 1000nL/min
Polystyrene/Divinylbenzene
Monolith Polymer
1 to 10
Analytical Column, NanoViper
Videos