missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepstatin A, Fisher BioReagents
£66.65 - £1120.16
Chemical Identifiers
CAS | 26305-03-3 |
---|---|
Molecular Formula | C34H63N5O9 |
Molecular Weight (g/mol) | 685.904 |
InChI Key | FAXGPCHRFPCXOO-WQILDUGRSA-N |
Synonym | pepstatin a |
PubChem CID | 131801262 |
IUPAC Name | (3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid |
SMILES | CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011413 SDS |
Fisher Bioreagents
BP2671-5 |
5mg | Amber Glass |
£66.65 5mg |
EU Stock 3 | Please sign in to purchase this item. Need a web account? Register with us today! | 10786834 SDS |
Fisher Bioreagents
BP2671-10 |
10mg | Amber Glass |
£87.41 10mg |
Estimated Shipment date from Supplier 07-01-2020 |
Please sign in to purchase this item. Need a web account? Register with us today! | 10264583 SDS |
Fisher Bioreagents
BP2671-25 |
25mg | Amber Glass |
£181.14 25mg |
EU Stock 2 | Please sign in to purchase this item. Need a web account? Register with us today! | 10410464 SDS |
Fisher Bioreagents
BP2671-100 |
100mg | Amber Glass |
£508.23 100mg |
EU Stock 2 | Please sign in to purchase this item. Need a web account? Register with us today! | 10051553 SDS |
Fisher Bioreagents
BP2671-250 |
250mg | Amber Glass |
£1120.16 250mg |
EU Stock 4 | Please sign in to purchase this item. Need a web account? Register with us today! |
Description
Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.Chemical Identifiers
26305-03-3 | |
685.904 | |
pepstatin a | |
(3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid |
C34H63N5O9 | |
FAXGPCHRFPCXOO-WQILDUGRSA-N | |
131801262 | |
CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O |
Specifications
235°C | |
Pepstatin A | |
Conforms to reference | |
Amber Glass | |
26305-03-3 | |
15, 7260 | |
FAXGPCHRFPCXOO-WQILDUGRSA-N | |
(3R,4S)-3-hydroxy-4-[[(2R)-2-[[(3R,4S)-3-hydroxy-6-methyl-4-[[(2S)-3-methyl-2-[[(2S)-3-methyl-2-(3-methylbutanoylamino)butanoyl]amino]butanoyl]amino]heptanoyl]amino]propanoyl]amino]-6-methylheptanoic acid | |
131801262 | |
Powder/Solid |
5mg | |
Pass Test | |
230° to 240°C | |
White | |
C34H63N5O9 | |
pepstatin a | |
CC(C)CC(C(CC(=O)O)O)NC(=O)C(C)NC(=O)CC(C(CC(C)C)NC(=O)C(C(C)C)NC(=O)C(C(C)C)NC(=O)CC(C)C)O | |
685.904 | |
685.9 |