missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pepstatin A, 98%
149.40 GBP valid until 2024-03-30
Use promo code "21648" to get your promotional price.
110.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21648" to get your promotional price.
110.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 mg
25 mg
Product Type:
Pepstatin
Protease Inhibitor
Description
Pepstatin A, is used as an irreversible inhibitor of HIV-1, aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d. Pepstatin A also efficiently inhibits HIV-1 protease and Fcγ receptor induction by IFN-γ.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Description | White to Off-white |
For Use With (Application) | Protease inhibitor that acts on pepsin, renin and cathepsin D |
Molecular Weight (g/mol) | 685.90g/mol, 685.90 |
Purity | 98% |
Molecular Formula | C34H63N5O9, C34H63N5O9 |
Content And Storage | 2°C to 8°C |
Beilstein | 2201362 |
Form | Powder |
InChI Key | FAXGPCHRFPCXOO-JHTFMCFMSA-N |
SMILES | CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'