missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Pepsin, porcine stomach

Brand:  Alfa Aesar™ J61679.09

Code : NEW

Additional Details : CAS Number : 9001-75-6 Weight : 0.01000kg

 View more versions of this product

Product Code. 15474229

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Powder
Pepsin
−30° to −10°C
EC 3.4.23.1; Pepsin A
White
10g
9001-75-6
Soluble in water and dilute aqueous buffers
Videos
SDS