Learn More
Pepsin, for biochemistry, powder, ACROS Organics™
Brand: Acros Organics 417070025
Code : 29
Additional Details : CAS Number : 9001-75-6 Weight : 2.55000kg
Quantity | 2.5kg |
---|---|
Packaging | Plastic Bottle |
Chemical Identifiers
Pepsin | |
9001-75-6 | |
MFCD00081840 |
For biochemistry, Powder | |
For biochemistry |
Specifications
Pepsin | |
For biochemistry, Powder | |
For biochemistry | |
MFCD00081840 | |
>50°C |
2.5kg | |
9001-75-6 | |
Plastic Bottle | |
15, 7259 |