missing translation for 'onlineSavingsMsg'
Learn More

PEPA, Tocris Bioscience™

Allosteric potentiator of AMPA receptor desensitization

Brand:  Tocris (Bio-Techne) 292910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462064

  • £117.45 / 10mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Novel allosteric potentiator of AMPA receptor desensitization. Slows the rate of onset of desensitization and potentiates steady-state equilibrium currents induced by glutamate with subunit (GluR3/4 > GluR1) and splice variant (flop > flip) selectivity. Ameliorates post-ischemic memory impairment in rats following i.v. administration.
Specifications

Chemical Identifiers

141286-78-4
402.431
pepa, 2,6-difluoro-4-2-phenylsulfonylamino ethylthio phenoxyacetamide, 2-2,6-difluoro-4-2-phenylsulfonyl amino ethyl sulfanyl phenoxy acetamide, lopac-d-8941, pepa drug, 4-2-phenylsulfonylamino ethylthio-2,6-difluorophenoxyacetamide, 4-2-phenylsulfonylamino ethylthio-2,6-difluoropheno-xyacetamide, 2,6-difluoro-4-2-phenylsul-fonyl-amino-ethyl-thio-phenoxy-acet-amide, 2,6-difluoro-4-2-phenylsul-fonyl-amino-ethyl-thio-phenoxy-acetamide, 2-2,6-difluoro-4-2-phenylsulfonyl amino ethyl thio phenoxy acetamide
2-[4-[2-(benzenesulfonamido)ethylsulfanyl]-2,6-difluorophenoxy]acetamide
C16H16F2N2O4S2
GTACSIONMHMRPD-UHFFFAOYSA-N
6603828
C1=CC=C(C=C1)S(=O)(=O)NCCSC2=CC(=C(C(=C2)F)OCC(=O)N)F

Specifications

PEPA
141286-78-4
pepa, 2,6-difluoro-4-2-phenylsulfonylamino ethylthio phenoxyacetamide, 2-2,6-difluoro-4-2-phenylsulfonyl amino ethyl sulfanyl phenoxy acetamide, lopac-d-8941, pepa drug, 4-2-phenylsulfonylamino ethylthio-2,6-difluorophenoxyacetamide, 4-2-phenylsulfonylamino ethylthio-2,6-difluoropheno-xyacetamide, 2,6-difluoro-4-2-phenylsul-fonyl-amino-ethyl-thio-phenoxy-acet-amide, 2,6-difluoro-4-2-phenylsul-fonyl-amino-ethyl-thio-phenoxy-acetamide, 2-2,6-difluoro-4-2-phenylsulfonyl amino ethyl thio phenoxy acetamide
C1=CC=C(C=C1)S(=O)(=O)NCCSC2=CC(=C(C(=C2)F)OCC(=O)N)F
402.431
402.44
10mg
C16H16F2N2O4S2
GTACSIONMHMRPD-UHFFFAOYSA-N
2-[4-[2-(benzenesulfonamido)ethylsulfanyl]-2,6-difluorophenoxy]acetamide
6603828
>99%
Videos