missing translation for 'onlineSavingsMsg'
Learn More

Pentyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-179822
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3954-13-0
C6H11NO
113.16
MFCD00144846
VRVUKQWNRPNACD-UHFFFAOYSA-N
pentyl isocyanate, pentylisocyanate, pentane, 1-isocyanato, 1-pentyl isocyanate, n-pentyl isocyanate, 1-isocyanato-pentane, amylisocyanat, n-pentylisocyanate, n-amyl isocyanate, acmc-1clhg
206036
1-isocyanatopentane
CCCCCN=C=O
This item is not returnable. View return policy
3954-13-0
C6H11NO
113.16
MFCD00144846
VRVUKQWNRPNACD-UHFFFAOYSA-N
pentyl isocyanate, pentylisocyanate, pentane, 1-isocyanato, 1-pentyl isocyanate, n-pentyl isocyanate, 1-isocyanato-pentane, amylisocyanat, n-pentylisocyanate, n-amyl isocyanate, acmc-1clhg
206036
1-isocyanatopentane
CCCCCN=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8700g/mL
Boiling Point 136.0°C to 137.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4NCO
Refractive Index 1.4130 to 1.4150
Quantity 1 g
Specific Gravity 0.87
Formula Weight 113.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Pentyl isocyanate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'