missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Pentraxin 2/SAP Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Pentraxin 2/SAP |
---|---|
Clone | 300103 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Mouse samples. It is validated for Western Blot.Specifications
Pentraxin 2/SAP | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P12246 | |
325 | |
Mouse myeloma cell line NS0-derived recombinant mouse Pentraxin 2/SAP Gln21-Asp224 Accession # P12246 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Pentraxin 2/SAP in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant rat Pentraxin 2/SAP is observed. In Western blots, 100% cross-reactivity with recombinant human (rh) Pentraxin 2/SAP is observed, but no cross-reactivity with rhPentraxin 2/SAP in direct ELISAs is observed. |
300103 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component | |
APCS | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only