missing translation for 'onlineSavingsMsg'
Learn More

Pentostatin, Tocris Bioscience™

Adenosine deaminase inhibitor

Brand:  Tocris (Bio-Techne) 203310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13439393

  • £132.30 / 10mg

Estimated Shipment date
from Supplier 01-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Irreversible inhibitor of adenosine deaminase (Ki = 2.5 pM). Anticancer agent.
Specifications

Chemical Identifiers

53910-25-1
268.273
(8R)-3-(2-Deoxy-β-D-erythro-pentofuranosyl)-3,4,7,8-tetrahydroimidazo[4,5-d][1,3]diazepin-8-ol
(8S)-3-[(2S,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-7,8-dihydro-4H-imidazo[4,5-d][1,3]diazepin-8-ol
C11H16N4O4
FPVKHBSQESCIEP-XAVMHZPKSA-N
97041463
C1C(C(OC1N2C=NC3=C2NC=NCC3O)CO)O

Specifications

Pentostatin
53910-25-1
(8R)-3-(2-Deoxy-β-D-erythro-pentofuranosyl)-3,4,7,8-tetrahydroimidazo[4,5-d][1,3]diazepin-8-ol
C1C(C(OC1N2C=NC3=C2NC=NCC3O)CO)O
268.273
268.27
10mg
C11H16N4O4
FPVKHBSQESCIEP-XAVMHZPKSA-N
(8S)-3-[(2S,4R,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-7,8-dihydro-4H-imidazo[4,5-d][1,3]diazepin-8-ol
97041463
>99%
Videos
Documents