missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™
49.81 GBP valid until 2024-03-31
Use promo code "21663" to get your promotional price.
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
6 x 1 L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Density | 0.626g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 0.083 |
Flash Point | −49°C (−56.2°F) |
Ignition Point | 500°F |
Linear Formula | CH3(CH2)3CH3 |
UN Number | UN1265 |
Beilstein | 969132 |
Formula Weight | 72.15g/mol |
Percent Purity | ≥99% (GC) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'