missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienylhafnium trichloride, 97%, ACROS Organics™

Brand:  Acros Organics 353490010

Code : 29

Additional Details : CAS Number : 75181-08-7 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10190362

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Pentamethylcyclopentadienylhafnium trichloride
97%
C10H15Cl3Hf
MFCD00070461
75181-08-7
97%
420.07

Specifications

Pentamethylcyclopentadienylhafnium trichloride
170.0° to 172.0°C
75181-08-7
Glass bottle
97%
C10H15Cl3Hf
MFCD00070461
Purple
1g
Authentic
Crystals or Crystalline Powder
97%
420.07
Videos
SDS