missing translation for 'onlineSavingsMsg'
Learn More

(Pentamethylcyclopentadienyl)iridium(III) Chloride dimer, 99%, Thermo Scientific Chemicals

Catalog Number p-3737149
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
12354-84-6
C20H30Cl4Ir2
796.73
MFCD00075435
MMAGMBCAIFVRGJ-UHFFFAOYSA-J
unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di
76030743
iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride
C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3]
12354-84-6
C20H30Cl4Ir2
796.73
MFCD00075435
MMAGMBCAIFVRGJ-UHFFFAOYSA-J
unii-6oy4uuc534, dichloro pentamethylcyclopentadienyl iridium iii dimer, pentamethylcyclopentadienyl iridium iii dichloride dimer, pentamethylcyclopentadienyliridium dichloride, pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer, eta5-pentamethylcyclopentadienyl iridium dichloride dimer, pentamethylcyclopentadienyliridium iii dichloride dimer mi, bis mu-chloro chloro eta5-pentamethylcyclopentadienyl iridium, iridium, di-mu-chlorodichlorobis 1,2,3,4,5-eta-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl di
76030743
iridium(3+);1,2,3,4,5-pentamethylcyclopenta-1,3-diene;tetrachloride
C[C-]1C(=C(C(=C1C)C)C)C.C[C-]1C(=C(C(=C1C)C)C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3].[Ir+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex Ir)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 7226
Formula Weight 796.73
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material (Pentamethylcyclopentadienyl)iridium(III) chloride dimer
Show More Show Less
compliance-icons
Product Identifier
  • (Pentamethylcyclopentadienyl)iridium(III) chloride dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'