missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Pentakis(dimethylamino)niobium(V), 99% (metals basis)

Preparation of niobium nitride films

Brand:  Alfa Aesar™ 042761.03

Code : NEW

Additional Details : CAS Number : 19824-58-9 Weight : 0.00100kg Transport : UN number : 3131 Chem class : 1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11375567

  • £86.71 / 1g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Pentakis (dimethylamino)niobium (V)
C10H30N5Nb
313.29
UN3131
19824-58-9
Nb[N(CH3)2]5
MFCD01863477
Niobium dimethylamide

Specifications

Pentakis (dimethylamino)niobium (V)
Moisture Sensitive
Dark purple
19824-58-9
99%
C10H30N5Nb
313.29
UN3131
Amine-like
100°C (0.1mmHg)
1g
Powder
(metals basis)
Nb[N(CH3)2]5
MFCD01863477
Niobium dimethylamide
Videos
SDS