Learn More
Alfa Aesar™ Pentakis(diethylamino)tantalum
Preparation of TaN films
Brand:  Alfa Aesar™ 042855.06
Code : NEW
Additional Details : CAS Number : 55161-66-5 Weight : 0.00500kg Transport : UN number : 3267 Chem class : 1b Pack group : II
Quantity 5g
 View more versions of this product
Product Code. 11360088
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Chemical Identifiers
Pentakis (diethylamino)tantalum
C20H50N5Ta
541.6
UN3267
55161-66-5
Ta[N(CH2CH3)2]5
MFCD01868625
Tantalum diethylamide
Specifications
Pentakis (diethylamino)tantalum
Liquid
Ta[N(CH2CH3)2]5
MFCD01868625
Tantalum diethylamide
5g
55161-66-5
C20H50N5Ta
541.6
UN3267
Orange
Moisture Sensitive