missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorothiophenol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Pentafluorothiophenol is used as a fluorinated building block
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentafluorothiophenol is used as a fluorinated building block
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Pentafluorothiophenol is used as a fluorinated building block
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | -24°C |
Density | 1.572 |
Boiling Point | 143°C |
Flash Point | 51°C (123°F) |
Odor | Stench |
Refractive Index | 1.465 |
Quantity | 5 g |
UN Number | UN2920 |
Beilstein | 1876292 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'