Learn More
Alfa Aesar™ (Pentafluorophenyl)diphenylphosphine, 98%
Brand: Alfa Aesar™ A11422.03
Code : NEW
Additional Details : CAS Number : 5525-95-1 Weight : 0.00100kg
Quantity | 1g |
---|
Description
ApplicationsThe reaction of (pentafluorophenyl)diphenylphosphine with C-ethoxycarbonyl-(p-methoxyphenyl) nitrile imine was used to synthesize [2,3,4,5-tetrafluoro-6-(p- methoxyphenylamino)phenyl]diphenylphosphine oxide whose reduction with trichlorosilane. the reactions of (pentafluorophenyl)diphenylphosphine and bis(penta- fluorophenyl)(phenyl)phosphine with nitrilimines (Housgen 1,3-dipoles) involves two-stage cycloaddi- tion and leads to formation of sufficiently stable fluorophosphoranes.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
(Pentafluorophenyl)diphenylphosphine | |
C18H10F5P | |
MFCD00000290 |
5525-95-1 | |
352.25 | |
Diphenylpentafluorophenylphosphine |
Specifications
(Pentafluorophenyl)diphenylphosphine | |
98% | |
C18H10F5P | |
352.25 | |
Diphenylpentafluorophenylphosphine |
5525-95-1 | |
70° to 72°C | |
1g | |
MFCD00000290 |