missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (Pentafluorophenyl)diphenylphosphine, 98%

Brand:  Alfa Aesar™ A11422.06

Code : NEW

Additional Details : CAS Number : 5525-95-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11490263

  • £300.90 / 5g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Applications
The reaction of (pentafluorophenyl)diphenylphosphine with C-ethoxycarbonyl-(p-methoxyphenyl) nitrile imine was used to synthesize [2,3,4,5-tetrafluoro-6-(p- methoxyphenylamino)phenyl]diphenylphosphine oxide whose reduction with trichlorosilane. the reactions of (pentafluorophenyl)diphenylphosphine and bis(penta- fluorophenyl)(phenyl)phosphine with nitrilimines (Housgen 1,3-dipoles) involves two-stage cycloaddi- tion and leads to formation of sufficiently stable fluorophosphoranes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

(Pentafluorophenyl)diphenylphosphine
C18H10F5P
MFCD00000290
5525-95-1
352.25
Diphenylpentafluorophenylphosphine

Specifications

(Pentafluorophenyl)diphenylphosphine
98%
C18H10F5P
352.25
Diphenylpentafluorophenylphosphine
5525-95-1
70° to 72°C
5g
MFCD00000290
Videos
SDS