missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 6-thien-2-ylnicotinate, 97%, Thermo Scientific™

Catalog Number p-3541159
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
926921-59-7
C16H6F5NO2S
371.28
MFCD09702374
UYAXHNWJTLZXTM-UHFFFAOYSA-N
pentafluorophenyl 2-thien-2-ylpyridine-5-carboxylate, pentafluorophenyl 6-thien-2-ylnicotinate, pentafluorophenyl 6-thiophen-2-yl pyridine-3-carboxylate, 2,3,4,5,6-pentafluorophenyl 6-thiophen-2-yl pyridine-3-carboxylate, pentafluorophenyl 6-thiophen-2-ylnicotinate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-thiophen-2-ylpyridine-3-carboxylate, 6-thiophen-2-yl-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
24229543
(2,3,4,5,6-pentafluorophenyl) 6-thiophen-2-ylpyridine-3-carboxylate
FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CS2)C(F)=C1F
This item is not returnable. View return policy
926921-59-7
C16H6F5NO2S
371.28
MFCD09702374
UYAXHNWJTLZXTM-UHFFFAOYSA-N
pentafluorophenyl 2-thien-2-ylpyridine-5-carboxylate, pentafluorophenyl 6-thien-2-ylnicotinate, pentafluorophenyl 6-thiophen-2-yl pyridine-3-carboxylate, 2,3,4,5,6-pentafluorophenyl 6-thiophen-2-yl pyridine-3-carboxylate, pentafluorophenyl 6-thiophen-2-ylnicotinate, 2,3,4,5,6-pentakis fluoranyl phenyl 6-thiophen-2-ylpyridine-3-carboxylate, 6-thiophen-2-yl-3-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
24229543
(2,3,4,5,6-pentafluorophenyl) 6-thiophen-2-ylpyridine-3-carboxylate
FC1=C(F)C(F)=C(OC(=O)C2=CN=C(C=C2)C2=CC=CS2)C(F)=C1F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 371.28
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 6-thien-2-ylnicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'