missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl 2-morpholinobenzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-ylbenzoate |
Formula Weight | 373.28 |
Percent Purity | 95% |
Chemical Name or Material | Pentafluorophenyl 2-morpholinobenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'