missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl 2-[4-(tert-butoxycarbonyl)piperazin-1-yl]isonicotinate, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 473.4 |
Percent Purity | 90% |
Chemical Name or Material | Pentafluorophenyl 2-[4-(tert-butoxycarbonyl)piperazin-1-yl]isonicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'