Learn More
Thermo Scientific Maybridge Pentafluorophenyl 1-methyl-1H-imidazole-4-carboxylate, 1g, 97%
Brand: Thermo Scientific Maybridge CC03626DA
Pentafluorophenyl 1-methyl-1H-imidazole-4-carboxylate, 1g, 97%
Code : 31
Additional Details : CAS Number : 898288-88-5 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III