missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals

Catalog Number p-7081836
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
827-15-6
C6F5I
293.962
MFCD00001032
OPYHNLNYCRZOGY-UHFFFAOYSA-N
iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
70008
1,2,3,4,5-pentafluoro-6-iodobenzene
C1(=C(C(=C(C(=C1F)F)I)F)F)F
This item is not returnable. View return policy
827-15-6
C6F5I
293.962
MFCD00001032
OPYHNLNYCRZOGY-UHFFFAOYSA-N
iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
70008
1,2,3,4,5-pentafluoro-6-iodobenzene
C1(=C(C(=C(C(=C1F)F)I)F)F)F

Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.

Solubility
Insoluble in water.

Notes
Light sensitive. Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -29°C
Density 2.212
Boiling Point 161°C to 163°C
Refractive Index 1.496
Quantity 5 g
Beilstein 2051549
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 293.96
Percent Purity 97%
Chemical Name or Material Pentafluoroiodobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'