missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 10523501
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2251-50-5
C7ClF5O
230.52
MFCD00000657
MYHOHFDYWMPGJY-UHFFFAOYSA-N
pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
75256
CHEBI:39425
FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F
This item is not returnable. View return policy
2251-50-5
C7ClF5O
230.52
MFCD00000657
MYHOHFDYWMPGJY-UHFFFAOYSA-N
pentafluorobenzoyl chloride, perfluorobenzoyl chloride, benzoyl chloride, pentafluoro, pentafluorobenzoylchloride, pubchem15522, acmc-1covi, intermediates-zcf02601, pentafluorobenzoic acid chloride, attercop-chm at133861, 67924-15-6 unspecified acetate
75256
CHEBI:39425
FC1=C(F)C(F)=C(C(Cl)=O)C(F)=C1F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.6000g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6F5COCl
Refractive Index 1.4520 to 1.4540
Quantity 25 g
Specific Gravity 1.6
Solubility Information Solubility in water: decomposes.
IUPAC Name 2,3,4,5,6-pentafluorobenzoyl chloride
Formula Weight 230.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Pentafluorobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Pentafluorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'