missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylenehexamine, Tech., 30.5% nitrogen, ACROS Organics™

Brand:  Acros Organics 320292500

Code : 29

Additional Details : CAS Number : 4067-16-7 Weight : 0.28750kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 250mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10752111

  • £20.80 / 250mL


Estimated Shipment
09-02-2018
Add to basket

Specifications

Chemical Identifiers

Pentaethylenehexamine
Technical Grade
C10H28N6
232.44
4067-16-7
≥30.5% (NITROGEN)
H2N(CH2CH2NH)4CH2CH2NH2
MFCD00008167

Specifications

Pentaethylenehexamine
175°C
Clear yellow
250mL
Authentic
1.5086 to 1.5106
0.95
Liquid
≥30.5% (NITROGEN)
H2N(CH2CH2NH)4CH2CH2NH2
MFCD00008167
0.9500g/mL
380.0°C
− 35.0°C
4067-16-7
Glass bottle
Solubility in water: immiscible with water.
Technical Grade
0.5% max. (K.F.)
C10H28N6
232.44
Videos
SDS