missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanolide, 98%, Thermo Scientific Chemicals

Catalog Number p-7081390
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
106-02-5
C15H28O2
240.387
MFCD00039667
FKUPPRZPSYCDRS-UHFFFAOYSA-N
cyclopentadecanolide, exaltolide, pentadecanolide, 15-pentadecanolide, pentalide, thibetolide, pentadecan-15-olide, pentadecalactone, muskalactone, muskolactone
235414
oxacyclohexadecan-2-one
C1CCCCCCCOC(=O)CCCCCC1
This item is not returnable. View return policy
106-02-5
C15H28O2
240.387
MFCD00039667
FKUPPRZPSYCDRS-UHFFFAOYSA-N
cyclopentadecanolide, exaltolide, pentadecanolide, 15-pentadecanolide, pentalide, thibetolide, pentadecan-15-olide, pentadecalactone, muskalactone, muskolactone
235414
oxacyclohexadecan-2-one
C1CCCCCCCOC(=O)CCCCCC1

Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

Solubility
Soluble in alcohol, dipropylene glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 33°C to 37°C
Density 0.918
Boiling Point 137°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.47
Quantity 25 g
Beilstein 143710
Merck Index 14,3910
Solubility Information Soluble in alcohol,dipropylene glycol. Insoluble in water.
Formula Weight 240.39
Percent Purity 98%
Chemical Name or Material Pentadecanolide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'