missing translation for 'onlineSavingsMsg'
Learn More

Pentacene 98%, ACROS Organics™

Brand:  Acros Organics 129790010

Code : 29

Additional Details : CAS Number : 135-48-8 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10751414

  • £192.40 / 1g


Estimated Shipment
18-06-2018
Add to basket

Specifications

Chemical Identifiers

Pentacene
135-48-8
98%
278.34
98%
≥97.5%
C22H14
MFCD00003710

Specifications

Pentacene
135-48-8
Fine Crystalline Powder
98%
278.34
Glass bottle
Dark blue, purple, or black
98%
Authentic
≥97.5%
C22H14
MFCD00003710
15, 7216
1g
Videos
SDS