missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Pentacarbonyliron, 99.5%

As a catalyst in organic reactions, and as carbonyl iron for high-frequency coils

Brand:  Alfa Aesar™ 031113.30

Code : NEW

Additional Details : CAS Number : 13463-40-6 Weight : 0.25000kg Transport : UN number : 1994 Chem class : 1 Pack group : I

Quantity 250g

 View more versions of this product

Product Code. 15461177

  • £81.09 / 250g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Pentacarbonyliron
Fe(CO)5
MFCD00011001
14,5099
13463-40-6
195.9
UN1994
Iron pentacarbonyl; Iron carbonyl

Specifications

Pentacarbonyliron
1.457g/mL
99.5%
195.9
UN1994
1.5196
Soluble in nickel tetracarbonyl and organic solvents; Insoluble in water
−20°C
250g
Pungent
13463-40-6
Liquid
Fe(CO)5
MFCD00011001
14,5099
Iron pentacarbonyl; Iron carbonyl
Red
103°C (200°C (dec.))
−15° (5°F)
Air & Light Sensitive
Videos
SDS