missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10750133
|
Macherey-Nagel™
718765 |
0.13 mm |
£23.25
Each |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
10439351
|
Macherey-Nagel™
718761 |
0.25 mm |
£23.25
Each |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
10269741
|
Macherey-Nagel™
718760 |
0.17 mm |
£23.25
Each |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
10546481
|
Macherey-Nagel™
718762 |
0.5 mm |
£23.25
Each |
Estimated Shipment: 17-04-2024
Log in to see stock. |
|||||
10556481
|
Macherey-Nagel™
718763 |
0.75 mm |
£23.25
Each |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||||
Specifications
1 m |