missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pectin

Catalog Number p-3779918
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
9000-69-5
MFCD00081838
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
9000-69-5
MFCD00081838

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Orange to Orange-Red
pH 3.0 to 3.8 (1% soln.)
Physical Form Powder
Ash acid-insoluble ash: < 1%
Merck Index 15, 7169
Quantity 250 g
Percent Purity 67 to 71% (degree of esterification)
Grade Practical
Infrared Spectrum Authentic
Loss on Drying 12% max. (105°C, 2 hrs)
Packaging Plastic bottle
Particle Size 2% Max. (>60 Mesh)
Viscosity 30.5 to 36.5
Chemical Name or Material Pectin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'