missing translation for 'onlineSavingsMsg'
Learn More

Manutan Pearl grey multi drawer central locking unit with five drawers

Brand:  Manutan 428T009N

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12811349

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

A range of steel drawer units, ideal for document storage. Non-locking or central locking units available. Fully extendable drawers with drawer stops.
  • Five drawers
  • Pearl grey
  • Locking unit
  • Dimensions (H x W x D): 330mm x 283mm x 400mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers