missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PDGF R beta Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | PDGF R beta |
---|---|
Clone | 182106 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Western Blot.Specifications
PDGF R beta | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P05622 | |
5159 | |
Mouse myeloma cell line NS0-derived recombinant mouse PDGF R beta Leu32-Lys530 Accession # P05622 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse PDGF R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 10-20% cross-reactivity with recombinant human PDGF R beta is observed, but no cross-reactivity with recombinant mouse (rm) PDGF R alpha , rmFlt-3, rmSCF R, or rmLymphotoxin beta R is observed. |
182106 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
PDGFRB | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only