missing translation for 'onlineSavingsMsg'
Learn More

PDE3B, clone: SMCP3B, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1973-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15217297

  • £181.95 / 100µg


Estimated Shipment
23-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: Cyclic nucleotide phosphodiesterases (PDE) catalyze the hydrolysis of cAMP and cGMP thereby regulating the effects of these second messengers. There are several families of PDE which share common structural organization. The PDE3 family has two members, 3A and 3B that are products of two separate genes. PDE3B (∽130 kDa) is expressed predominantly in adipose tissue and lymphocytes, while PDE3A (∽120 kDa) is expressed predominantly in platelets and heart tissue. The monoclonal antibody SMCP3B reacts with mouse PDE3B. The SMCP3B antibody was generated against amino acids 643-1100 in the C-terminus and does not cross-react with mouse PDE3A, as determined by western blot analysis of lysates of mouse heart tissue. The SMCP3B antibody does not cross-react with human PDE3B.Applications Reported: This SMCP3B antibody has been reported for use in immunoblotting (WB).Applications Tested: This SMCP3B antibody has been tested by immunoblotting of mouse lymph node cells and mouse heart under reducing conditions. It can be used at 1-5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

PDE3B
SMCP3B
Unconjugated
Purified
Rat
100μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Phosphodiesterase 3b
IgG2a
RUO
Primary
Murine
Videos