missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PDE2A Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding amino acids (Glu 215-His 900) of human PDE2A fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 13031H08B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15256115

  • £314.39 / 50µg


Estimated Shipment
09-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
5138
≥85% by SDS-PAGE
Human
<1 EU/μg
PDE2A
Other Proteins
PDE2A Recombinant Human Protein
Lyophilized
80kd
50μg
Recombinant, Truncated
Phosphodiesterase 2A,cGMP-Stimulated
Assay Standard
Insect
His Tag
Videos