missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ PDCD1 Recombinant Mouse Protein, His Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 167) of mouse PDCD1 precursor fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 50124M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15312766

  • £123.88 / 25µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
18566
≥90% by SDS-PAGE
Mouse
<1 EU/μg
PDCD1
Other Proteins
PDCD1 Recombinant Mouse Protein
Lyophilized
17.6kd
5 x 5μg
Truncated, Recombinant
Programmed Cell Death 1
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.