Learn More
PDCD1 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™
Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 167) of mouse PDCD1 precursor fused to polyhistidine tag at C-terminus
Brand: Invitrogen 50124M08H50
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 167) of mouse PDCD1 precursor fused to polyhistidine tag at C-terminus
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications
Protein Structure Analysis, Protein Standard, Cell Biology and Analysis | |
17.6kd | |
Mouse | |
<1 EU/μg | |
PDCD1 | |
His Tag | |
Other Proteins | |
≥90% by SDS-PAGE |
18566 | |
50μg | |
Truncated, Recombinant | |
Programmed Cell Death 1 | |
PDCD1 Recombinant Mouse Protein | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.