Learn More
Cytiva PD MultiTrap™ Plates
Convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr) with high reproducibility from well to well and from plate to plate
Brand: Cytiva 28-9180-06
Code : GE
Additional Details : Weight : 3.30210kg
Description
- Designed for high-throughput desalting and buffer exchange of multiple samples
- Range of other applications include buffer exchange and removal of low-molecular weight compounds
- 96-well plates prepacked with 500μL Sephadex™ G-25 Medium/well for high reproducibility
- Suitable for automated use with automated robotic systems as well as manual use
- Sample preparation in the range of 70 to 130μL without sample dilution
- Stable in the pH range 2 to 13, and in the presence of all commonly used buffers
Specifications
0.2M NaOH, 0.2M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS , 24% Ethanol, 30% Porpanol, 30% Aceto nitrile | |
0.07 to 0.13mL | |
11.81 in. | |
>100cm/hr. | |
2 to 13 | |
Cross-linked Dextran | |
High-throughput desalting and Buffer exchange of multiple samples, use manually or with robotic systems |
PD Sephadex G-25 MultiTrap Plate | |
4 x 96 Well Plate | |
300mm | |
Polypropylene, Polyethylene | |
50μm in dry form, 38 to 235μm in wet form | |
4° to 30°C |
Safety and Handling
- PD MultiTrap G-25
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.