Learn More
Tocris Bioscience™ PD 98059
MEK inhibitor
Brand:  Tocris Bioscience™ 12131
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13488893
  • £73.40 / 1mg
Estimated Shipment
27-07-2018
Add to basket
Description
Description
Inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal. Also available as part of the MAPK Cascade Inhibitor Tocriset™ and MAPK Inhibitor Tocriset™ .
Specifications
Specifications
PD 98059
>98
1mg
PD 98059
267.28