missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD 407824, Tocris Bioscience™
Selective inhibitor of Chk1 and Wee1
Brand: Tocris (Bio-Techne) 269410
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
Selective inhibitor of checkpoint kinases Chk1 and Wee1 (IC50 values are 47 and 97 nM respectively). Displays selectivity over a range of other protein kinases; IC50 values are 3.4, 3.75, > 5, > 50, > 50 and > 50 μM for PKC, CDK4, other CDKs, c-Src, PDGFR and FGFR respectively.Chemical Identifiers
622864-54-4 | |
328.327 | |
9-hydroxy-4-phenyl-6h-pyrrolo 3,4-c carbazole-1,3-dione, 9-hydroxy-4-phenylpyrrolo 3,4-c carbazole-1,3 2h,6h-dione, 9-hydroxy-4-phenyl-2h,6h-pyrrolo 3,4-c carbazole-1,3-dione, pd hplc, 9-hydroxy-4-phenyl-pyrrolo 3,4-c carbazole-1,3 2h,6h-dione, pyrrolo 3,4-c carbazole-1,3 2h,6h-dione, 9-hydroxy-4-phenyl, 14-hydroxy-7-phenyl-4,10-diazatetracyclo 7.7.0.0^ 2,6 .0^ 11,16 hexadeca-1,6,8,11,13,15-hexaene-3,5-dione | |
9-hydroxy-4-phenyl-6H-pyrrolo[3,4-c]carbazole-1,3-dione |
C20H12N2O3 | |
IAUZTOZLTFSMIE-UHFFFAOYSA-N | |
4369491 | |
C1=CC=C(C=C1)C2=CC3=C(C4=C(N3)C=CC(=C4)O)C5=C2C(=O)NC5=O |
Specifications
PD 407824 | |
622864-54-4 | |
9-hydroxy-4-phenyl-6h-pyrrolo 3,4-c carbazole-1,3-dione, 9-hydroxy-4-phenylpyrrolo 3,4-c carbazole-1,3 2h,6h-dione, 9-hydroxy-4-phenyl-2h,6h-pyrrolo 3,4-c carbazole-1,3-dione, pd hplc, 9-hydroxy-4-phenyl-pyrrolo 3,4-c carbazole-1,3 2h,6h-dione, pyrrolo 3,4-c carbazole-1,3 2h,6h-dione, 9-hydroxy-4-phenyl, 14-hydroxy-7-phenyl-4,10-diazatetracyclo 7.7.0.0^ 2,6 .0^ 11,16 hexadeca-1,6,8,11,13,15-hexaene-3,5-dione | |
C1=CC=C(C=C1)C2=CC3=C(C4=C(N3)C=CC(=C4)O)C5=C2C(=O)NC5=O | |
328.327 | |
328.32 |
10mg | |
C20H12N2O3 | |
IAUZTOZLTFSMIE-UHFFFAOYSA-N | |
9-hydroxy-4-phenyl-6H-pyrrolo[3,4-c]carbazole-1,3-dione | |
4369491 | |
>98% |