Learn More
PD 166793, Tocris Bioscience™
Broad spectrum MMP inhibitor
POA
Description
Description
Broad spectrum MMP inhibitor. Displays high affinity for MMP-2, -3 and -13 (IC50 values are 4, 7 and 8 nM respectively) and exhibits > 750-fold selectivity over MMP-1, -7 and -9. Attenuates left ventricular remodelling and dysfunction in rat model of heart failure.