missing translation for 'onlineSavingsMsg'
Learn More

PD 166793, Tocris Bioscience™

Broad spectrum MMP inhibitor

Brand:  Tocris (Bio-Techne) 252050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461384

  • £378.85 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Broad spectrum MMP inhibitor. Displays high affinity for MMP-2, -3 and -13 (IC50 values are 4, 7 and 8 nM respectively) and exhibits > 750-fold selectivity over MMP-1, -7 and -9. Attenuates left ventricular remodelling and dysfunction in rat model of heart failure.
Specifications

Chemical Identifiers

199850-67-4
412.298
s-2-4'-bromo-1,1'-biphenyl-4-ylsulfonamido-3-methylbutanoic acid, unii-u855l3soxd, u855l3soxd, chembl43152, s-2-4'-bromobiphenyl-4-ylsulfonamido-3-methylbutanoic acid, mmp-2/mmp-3 inhibitor iii, pd166793, s-2-4′-bromo-biphenyl-4-sulfonylamino-3-methylbutyric acid, 2s-2-4'-bromo-1,1'-biphenyl-4-sulfonamido-3-methylbutanoic acid, l-valine, n-4'-bromo 1,1'-biphenyl-4-yl sulfonyl, alpha-arylsulfonylamino carboxylate 2s
(2S)-2-[[4-(4-bromophenyl)phenyl]sulfonylamino]-3-methylbutanoic acid
C17H18BrNO4S
GJOCABIDMCKCEG-INIZCTEOSA-N
9887870
CC(C)C(C(=O)O)NS(=O)(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)Br

Specifications

PD 166793
C17H18BrNO4S
GJOCABIDMCKCEG-INIZCTEOSA-N
(2S)-2-[[4-(4-bromophenyl)phenyl]sulfonylamino]-3-methylbutanoic acid
9887870
>99%
199850-67-4
s-2-4'-bromo-1,1'-biphenyl-4-ylsulfonamido-3-methylbutanoic acid, unii-u855l3soxd, u855l3soxd, chembl43152, s-2-4'-bromobiphenyl-4-ylsulfonamido-3-methylbutanoic acid, mmp-2/mmp-3 inhibitor iii, pd166793, s-2-4′-bromo-biphenyl-4-sulfonylamino-3-methylbutyric acid, 2s-2-4'-bromo-1,1'-biphenyl-4-sulfonamido-3-methylbutanoic acid, l-valine, n-4'-bromo 1,1'-biphenyl-4-yl sulfonyl, alpha-arylsulfonylamino carboxylate 2s
CC(C)C(C(=O)O)NS(=O)(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)Br
412.298
412.3
50mg
Videos