missing translation for 'onlineSavingsMsg'
Learn More

PD 153035 hydrochloride, Tocris Bioscience™

EGFR-kinase inhibitor

Brand:  Tocris (Bio-Techne) 1037/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458353

  • £137.70 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

An extremely potent inhibitor of epidermal growth factor (EGF) receptor tyrosine kinase, with an IC50 of 25 pM. Inhibits other purified tyrosine kinases only at micromolar or higher concentrations.
Specifications

Chemical Identifiers

183322-45-4
396.669
pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, pd153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride
N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride
C16H15BrClN3O2
ZJOKWAWPAPMNIM-UHFFFAOYSA-N
11246488
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl

Specifications

PD 153035 hydrochloride
183322-45-4
pd153035 hydrochloride, pd 153035 hydrochloride, n-3-bromophenyl-6,7-dimethoxyquinazolin-4-amine hydrochloride, pd-153035 hydrochloride, pd153035 hydrochloride, tyrphostin ag 1517, ag 1517 hydrochloride, su-5271 hydrochloride, unii-ahj252p69n, 4-3-bromophenyl amino-6,7-dimethoxyquinazoline hydrochloride
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=CC(=CC=C3)Br)OC.Cl
396.669
396.67
10mg
C16H15BrClN3O2
ZJOKWAWPAPMNIM-UHFFFAOYSA-N
N-(3-bromophenyl)-6,7-dimethoxyquinazolin-4-amine;hydrochloride
11246488
>98%
Videos