Learn More
PD 150606, Tocris Bioscience™
Cell permeable calpain inhibitor
Brand:  Tocris (Bio-Techne) 126910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13489023
  • £147.15 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Selective, cell-permeable non-peptide calpain inhibitor (Ki values for v and m-calpains are 0.21 and 0.37μM respectively). Targets the calcium binding sites of calpain. Demonstrates high specificity for calpains relative to other proteases, and is a non-competitive inhibitor with respect to the substrate.
Specifications
Chemical Identifiers
179528-45-1
306.117
(Z)-3-(4-Iodophenyl)-2-mercapto-2-propenoic acid
C9H7IO2S
DJCVSFWGKYHMKH-YVMONPNESA-N
9839500
Specifications
PD 150606
>95%
(Z)-3-(4-Iodophenyl)-2-mercapto-2-propenoic acid
DJCVSFWGKYHMKH-YVMONPNESA-N
(Z)-3-(4-iodophenyl)-2-sulfanylprop-2-enoic acid
9839500
179528-45-1
C9H7IO2S
10mg
C1=CC(=CC=C1C=C(C(=O)O)S)I
306.117
306.12
Videos