Learn More
(+)-PD 128907 hydrochloride, Tocris Bioscience™
D3 agonist (D3 ≥ D2 > D4)
Brand:  Tocris (Bio-Techne) 124350
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488953
  • £914.00 / 50mg
Estimated Shipment
07-03-2018
Add to basket
Description
Description
Potent D3 dopamine receptor agonist (Ki = 2.3 nM). Displays 18 - 200-fold selectivity over other dopamine receptor subtypes.
Specifications
Chemical Identifiers
(+)-PD 128907 hydrochloride
C14H19NO3·HCl
285.77
(4aR,10bR)-3,4a,4,10b-Tetrahydro-4-propyl-2H,5H-[1]benzopyrano-[4,3-b]-1,4-oxazin-9-ol hydrochloride
Specifications
(+)-PD 128907 hydrochloride
300576-59-4
C14H19NO3·HCl
(4aR,10bR)-3,4a,4,10b-Tetrahydro-4-propyl-2H,5H-[1]benzopyrano-[4,3-b]-1,4-oxazin-9-ol hydrochloride
50mg
>99%
285.77
Videos