missing translation for 'onlineSavingsMsg'
Learn More

PD 102807, Tocris Bioscience™

Selective M4 antagonist

Brand:  Tocris (Bio-Techne) 167150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13449893

  • £792.33 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective M4 muscarinic receptor antagonist. IC50 values are 91, 6559, 3441, 950 and 7412 nM for human M4, M1, M2, M3, and M5 receptors respectively.
Specifications

Chemical Identifiers

23062-91-1
392.455
chembl59898, 3,6a,11,14-tetrahydro-9-methoxy-2-methyl-12h-isoquino 1,2-b pyrrolo 3,2-f 1,3 benzoxazine-1-carboxylic acid, ethyl ester, 3,6a,11,14-tetrahydro-9-methoxy-2-methyl-12h-isoquino 1,2-b pyrrolo 3,2-f 1,3 benzoxazine-1-carboxylic acid ethyl ester, 9-methoxy-2-methyl-11,12-dihydro-3h,6ah,13h-6-oxa-3,12a-diaza-benzo a cyclopenta h anthracene-1-carboxylic acid ethyl ester
CCOC(=O)C1=C(NC2=C1C3=C(C=C2)OC4C5=C(CCN4C3)C=C(C=C5)OC)C
C23H24N2O4
VDDUJINYXKGZLV-UHFFFAOYSA-N
4995951

Specifications

50mg
23062-91-1
chembl59898, 3,6a,11,14-tetrahydro-9-methoxy-2-methyl-12h-isoquino 1,2-b pyrrolo 3,2-f 1,3 benzoxazine-1-carboxylic acid, ethyl ester, 3,6a,11,14-tetrahydro-9-methoxy-2-methyl-12h-isoquino 1,2-b pyrrolo 3,2-f 1,3 benzoxazine-1-carboxylic acid ethyl ester, 9-methoxy-2-methyl-11,12-dihydro-3h,6ah,13h-6-oxa-3,12a-diaza-benzo a cyclopenta h anthracene-1-carboxylic acid ethyl ester
CCOC(=O)C1=C(NC2=C1C3=C(C=C2)OC4C5=C(CCN4C3)C=C(C=C5)OC)C
4995951
>98%
PD 102807
C23H24N2O4
VDDUJINYXKGZLV-UHFFFAOYSA-N
392.455
392.45
Videos
missing translation for 'documents'